Learn More
Cliawaived Inc CLIAIwaived, Inc. 14 Panel Multi Drug Screen Cups
SDP
Supplier:  Cliawaived Inc CLIAMDSC14
Encompass_Preferred
(25 Cups per Box)Rapid drug test cup for detection of drugs of abuse in urine. Read results between 5-60 minutes. This cup is FDA Cleared, CLIA waived, and CE marked for export.Requires only 5 mL of urine to activate. Low sample volume saves $$ on use of Quality Controls. We will price match any better offer!The cup tests AMP 500, BAR300, BUP10, BZO300, COC150, EDDP300, MDMA500, MET500, MTD300, OPI300, OXY100, PCP25, TCA 1000, THC50
Catalog No. NC1859616