Learn More
MOVEX INC 14"""" Diameter Clear Polypropyline Hood with Connecting Tube
Supplier:  MOVEX INC MEK35075
Encompass_Preferred
The MEK dome hood is a clear dome hood that is suitable for lighter gases with a wider dispersal of contaminants without blocking users vision. Comes with hoodtube that will fit arm Ø75.
Catalog No. NC1835408