Learn More
Cayman Chemical 14 S-HDHA-d5 100UG
SDP
Supplier:  Cayman Chemical 9002764100 UG
Encompass_Preferred
An internal standard for the quantification of 14(S)-HDHA by GC- or LC-MS
Catalog No. 50-289-4741