Learn More
Medchemexpress LLC Δ14-Triamcinolone acetonide | 1260149-96-9 | 432.48 | 1 MG
SDP
Supplier:  Medchemexpress LLC 1MGHYZ8648
Encompass_Preferred
Δ14-Triamcinolone acetonide is identified as a potential impurity. It is known to inhibit basic fibroblast growth factor (bFGF) induced proliferation of retinal endothelial cells. This compound also contributes to the reduction of chondrocyte viability, which can lead to cartilage destruction. Furthermore, Δ14-Triamcinolone acetonide has applications in the study of various diseases, including atopic dermatitis.
  • Potential impurity.
  • Inhibits basic fibroblast growth factor (bFGF) induced proliferation of retinal endothelial cells.
  • Reduces chondrocyte viability and leads to cartilage destruction.
  • Can be used in the study of diseases such as atopic dermatitis.
Catalog No. 50-003-65677