Learn More
Cayman Chemical 14 ZEicosenoic Acid
SDP
Supplier:  Cayman Chemical 100093751 MG
Encompass_Preferred
14(Z)-Eicosenoic acid is a monounsaturated fatty acid. It is found in small quantities in marine life including the sponge (Tedania dirhaphis) and goldfish (Carassius auratus).{14592,14593} Potentially it could be the product of fatty acid chain elongation of 12(Z)-oleic acid.
Catalog No. 50-288-9077