Learn More
TARGETMOL CHEMICALS INC 1400W DIHYDROCHLORIDE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T349125MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1400W dihydrochloride (N-(3-(Aminomethyl)benzyl)acetamidine) is a highly effective and specific inhibitor of inducible NOS2 (iNOS). purity: 99%
Catalog No. 50-257-6933