Learn More
Fine Science Tools Mayo-Noble Scissors, Stainless Steel, 16.5 cm
SDP
Supplier:  Fine Science Tools 1401417
Encompass
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 37mm/ Alloy / Material: Stainless Steel/ Length: 16.5cm/ Serrated: No
Catalog No. NC0155090