Learn More
Sino Biological Recombinant Human IVD Protein (aa 30-423, His Tag)
Supplier:  Sino Biological 14206H07B100
Encompass_Preferred
A DNA sequence encoding the human IVD (AAH17202.1) (His30-His423) was fused with a polyhistide tag at the N-terminus.
Catalog No. 50-161-9040