Learn More
Sino Biological Recombinant Human ENSA Protein (His Tag)
Supplier:  Sino Biological 14321H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human ENSA (O43768-1) (Met1-Glu121) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-9100