Learn More
Sino Biological Recombinant Human CGB5 Protein (mFc Tag)
Supplier:  Sino Biological 14574H05H100
Encompass_Preferred
A DNA sequence encoding the human CGB5 (NP_000728.1) (Met1-Gln165) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-9244