Learn More
Sino Biological Recombinant Human EEF1B2 Protein (His Tag)
Supplier:  Sino Biological 14611H07E100
Encompass_Preferred
A DNA sequence encoding the human EEF1B2 (P24534)(Met1-Ile225) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9261