Learn More
Sino Biological Recombinant Human PSME2 Protein (His Tag)
Supplier:  Sino Biological 14640H07E100
Encompass_Preferred
A DNA sequence encoding the human PSME2 (Met1-Tyr239) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9285