Learn More
Sino Biological Recombinant Human Alpha SNAP Protein (His Tag)
Supplier:  Sino Biological 14800H07E100
Encompass_Preferred
A DNA sequence encoding the human NAPA (NP_003818.2) (Met1-Arg295) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9325