Learn More
Cayman Chemical 14deoxy11 12dIdehydro ndr 5mg
SDP
Supplier:  Cayman Chemical 116705 MG
Encompass_Preferred
A noncytotoxic analog of the natural diterpenoid that possesses anti-inflammatory, antiallergenic, immuno-stimulatory, antiviral, antioxidant, hepatoprotective, and cardiovascular activities
Catalog No. 50-295-2241