Learn More
Bellco Glass, Inc. 14mL Culture Tube and Dual Cap, PP, Sterile
SDP
Supplier:  Bellco Glass, Inc. 230439000
Encompass_Preferred
Dual caps allow for either vented or sealed positions14mL tubes feature molded graduationsPolystyrene tubes are optically clear to visualize culturesPolypropylene tubes offer better chemical resistance Pack Description25/Re-Sealable BagCase Quantity500SterileYesNon-PyrogenicYesRNase/DNase FreeYesAutoclavableYesPrimary ColorTube: NaturalSecondary ColorCap: NaturalPrimary MaterialTube: PolypropyleneSecondary MaterialCap: LDPEMaximum Volume14mLGraduatedYesGraduation Increments (Subdivisions)0.5mLMax Force (RCF) g3000 xg
Catalog No. 50-233-0332