Learn More
Cayman Chemical 14R 15S-EET 100UG
SDP
Supplier:  Cayman Chemical 9004351100 UG
Encompass_Preferred
An oxylipin and arachidonic acid metabolite; induces relaxation of precontracted isolated bovine coronary arteries
Catalog No. 50-289-5834