Learn More
MSC HAZRD MATRL CLASS SIGN
Supplier:  MSC 63377139
Encompass
Chemical & Hazardous Material Sign: Rectangle, "HAZARDOUS MATERIALS CLASSIFICATION HEALTH HAZARD 3 2 1 W 4-Deadly 3-Extreme danger 2-Hazardous 1-Slightly hazardous 0-Normal material FIRE HAZARD Flash Points 4-Below 73B: F 3-Below 100B: F 2-Below 200B: F 1
Catalog No. 50-273-7624