Learn More
Uline Industrial Poly Bags | | 250
Supplier:  Uline S3164
Encompass_Preferred
Heavier protection for industrial parts and tools, 100% virgin high clarity polyethylene film, Meets FDA and USDA specifications for food contact, Low/Linear Low Density, Sulfur free, Amine and Silicone free, Latex free, BHT free (Butylated Hydroxy Toluene), Chlorine free, PVC free, Temperature range 20 - 150 F, Recyclable 4, Bottom seal, Not treated for printing by end-user, Not recommended for dry ice, Not microwaveable, Cannot be used for baking, cooking, or boiling, Do not store in sub-freezing temperatures, 3 or 4 mil is recommended for use in a trash compactor, Water-resistant, not waterproof
Catalog No. NC2391364