Learn More
MAGIC BIOPROCESSING MAGic™Accio Pilot 50
Supplier:  MAGIC BIOPROCESSING 2200
Encompass_Preferred
MAGic™Accio PILOT is designed to support development-phase and medium-scale purification of nucleic acids, peptides, proteins, antibodies, and viruses from both clarified and unclarified cell culture mixtures. Capable of processing volumes from 5 mL to several liters, it works seamlessly with 20 µL to 250 mL of settled MAGic™ Beads. Used with the MAGic™Accio PILOT 50, 500, or 2000 units, it enables rapid, durable manual separation in both serial and parallel configurations. By combining units, users can isolate low-expressing targets from large volumes and concentrate them into smaller vessels, streamlining workflows and boosting yield.
Catalog No. NC3724017