Learn More
Cayman Chemical 15 6nItroxyhexnoyl17phe 10mg
SDP
Supplier:  Cayman Chemical 3279110 MG
Encompass_Preferred
A nitric oxide-donating derivative of 17-phenyl trinor PGF2α; reduces IOP in a rabbit model of hypertonic saline-induced transient ocular hypertension at 0.14%; reduces IOP in a cynomolgus monkey model of laser-induced ocular hypertension at 0.042%
Catalog No. 50-294-7538