Learn More
Medchemexpress LLC 15-Azido-pentadecanoic acid | 118162-46-2 | C15H29N3O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1516565MG
Encompass_Preferred
15-Azido-pentadecanoic acid is a click chemistry reagent featuring an azide group. This reagent is utilized for identifying and characterizing post-translationally palmitylated proteins through a two-step labeling and detection process.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne-containing molecules.
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Suitable for research use only.
Catalog No. 50-003-73997