Learn More
Medchemexpress LLC 15-Azido-pentadecanoic acid (Azido Palmitic Acid) | 118162-46-2 | C15H29N3O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15165650MG
Encompass_Preferred
15-Azido-pentadecanoic acid is a click chemistry reagent containing an azide group. It can be used to identify and characterize post-translationally palmitylated proteins using a simple and robust two-step labeling and detection technique.
  • Contains an azide group for click chemistry reactions
  • Compatible with SPAAC reactions using DBCO or BCN groups
  • Identifies and characterizes post-translationally palmitylated proteins
Catalog No. 50-003-72944