Learn More
Cayman Chemical 15-keto Fluprostenol 5mg
SDP
Supplier:  Cayman Chemical 167855 MG
Encompass_Preferred
A derivative of fluprostenol; a potential inactive metabolite of 15-keto fluprostenol isopropyl ester; a potential metabolite of fluprostenol (travoprost); a potential impurity found in commercial preparations of fluprostenol
Catalog No. 50-296-5742