Learn More
Chemglass Life Sciences Chemglass Tripod Support Stand for 15 L Unjacketed Bioreactor (CLS-1385-STD15)
SDP
Supplier:  Chemglass Life Sciences CLS1385STD15
Encompass
Chemglass tripod support stand designed to securely hold 15 L unjacketed bioreactor vessels. The complete stand provides a stable mounting platform for the vessel headplate and includes the attachment hardware needed for routine setup and operation.
  • Designed for 15 L unjacketed bioreactor vessels
  • Top two-piece ring made of anodized aluminum
  • Lower ring and legs made of electropolished stainless steel
  • Includes silicone o-ring and six knurled nuts for headplate attachment
  • Compatible with optional stabilizer bars and replacement o-rings
Catalog No. NC2832415