Learn More
Polycarbin 15 mL Centrifuge Tube CS500, Bagged (Sterile)
SDP
Supplier:  Polycarbin PC020201G
Encompass_Preferred
Polycarbin's bulk 15 mL conical tubes are made using 20% circular economy, post-consumer recycled content. Each lot of tubes is sterile. Certified DNase, RNase, and Pyrogen free. 50 tubes/rack, 10 racks/case, 500 tubes/case. Centrifugation Speed 12,000 x g. Temperature Range of -196°C to 100°C. Manufactured in a ISO 9001 compliant facility.
Catalog No. NC2603179