Learn More
Medchemexpress LLC 15-PGDH-IN-1 | 2241676-74-2 | C24H22N4O2 | 5 MG

Supplier: Medchemexpress LLC HY1518075MG
15-PGDH-IN-1 is a potent and orally active 15-PGDH inhibitor. It demonstrates inhibition activity against recombinant human 15-PGDH with an IC50 value of 3 nM and is utilized for research into tissue repair and regeneration.
- Potent and orally active 15-PGDH inhibitor
- Inhibits recombinant human 15-PGDH with an IC50 value of 3 nM
- Used for research of tissue repair and regeneration
- Molecular weight: 398.46
- Purity: 98.35%
- Appearance: Solid, light yellow to yellow
- Storage conditions: Powder at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.