Learn More
Cayman Chemical 15 R-17-phenyl trinor Prosta
SDP
Supplier:  Cayman Chemical 1681410 MG
Encompass_Preferred
A 15(R) isomer of 17-phenyl trinor PGF2α and active metabolite of 17-phenyl trinor PGF2α ethyl amide; potential agonist of the FP receptor; induces relaxation of isolated cat iris sphincter muscles (EC50 = 30 nM)
Catalog No. 50-296-5648