Learn More
Cayman Chemical 15 R-17-phenyl trinor Prosta
SDP
Supplier:  Cayman Chemical 1682550 MG
Encompass_Preferred
A latanoprost-related isomer containing both a double bond at C-13,14 and an invered (b) hydroxyl group at C-15; a potential impurity in most commercial preparations of the lantanoprost bulk drug product
Catalog No. 50-296-6096