Learn More
Cayman Chemical 15 RPinane Thromboxane A2
SDP
Supplier:  Cayman Chemical 100085101 MG
Encompass_Preferred
An epimer of PTA2; does not inhibit collagen-induced platelet aggregation (IC50s = 120-130 µM); does not affect gastric tone in isolated rat gastric fundus when used at concentrations of 0.5 or 1.5 UG/ml; less effective than PTA2 at inhibiting prostaglandin-induced contraction of isolated rat stomach muscle
Catalog No. 50-289-0085