Learn More
Cayman Chemical 15 S-HEPE-d5 ethyl ester 50ug
SDP
Supplier:  Cayman Chemical 3378350 UG
Encompass_Preferred
An internal standard for the quantification of 15-HEPE by GC- or LC-MS
Catalog No. 50-295-0361