Learn More
Cayman Chemical 15 S-Latnoprost 10mg
SDP
Supplier:  Cayman Chemical 1681810 MG
Encompass_Preferred
A 15(S) isomer of latanoprost; reduces IOP in cynomolgus monkeys and does not induce eye irritation in cats when applied topically at 3 UG/eye; a potential impurity in commercial preparations of latanoprost
Catalog No. 50-296-7634