Learn More
Chemglass Life Sciences 150 mL Buchner Funnel With Fine Fritted Disc
SDP
Supplier:  Chemglass Life Sciences CG8590150F
Encompass_Preferred
A 150 mL borosilicate glass Buchner funnel with a sealed-in fine fritted glass disc for chemical-resistant filtration. The fritted disc provides a reusable filtration surface; do not subject the disc to a pressure differential greater than 1 atmosphere. The top edge is beaded for handling.
  • Sealed-in fine fritted glass disc for reusable filtration.
  • Suitable when filter paper cannot withstand chemical attack.
  • Borosilicate glass construction for chemical resistance.
  • Beaded top edge for secure handling.
  • Not rated for pressure differentials greater than 1 atmosphere across the frit.
Catalog No. NC3706699