Learn More
Chemglass Life Sciences Reaction Vessel Lid, 3-Neck, 29/42 Center Neck, 2-29/42 Side Necks, 150mm Schott Flange
SDP
Supplier:  Chemglass Life Sciences CG194005
Encompass_Preferred
Reaction Vessel Lid, 3-Neck, 29/42 Center Neck, 2-29/42 Side Necks, 150mm Schott Flange. Three neck vessel lid to fit CG-1920 through CG-1936 vessels. Lid has a flat ground surface allowing it to be greased or used with an o-ring. NOTE: Three neck lids are available with joint sizes and configurations other than those listed, please call for details.
Catalog No. CG194005