Learn More
Malvern Panalytical 150Nm/15Ml Polys'Rene Sta
Supplier:  Malvern Panalytical LTX3150A
Encompass
Nanosphere Size Standards 150nm /15ml. “Nanosphere Size Standards are highly uniform polystyrene spheres calibrated by NIST traceable standards and packaged as aqueous suspensions in dropper-tipped bottles. 1% solids, optimized for easy dispersal and colloidal stability. Includes Certificate of Calibration and Traceability to NIST.” 
Catalog No. NC1684608