Learn More
Sino Biological Recombinant Human CNKSR3 Protein (GST Tag)
Supplier:  Sino Biological 15188H09B100
Encompass_Preferred
A DNA sequence encoding the human CNKSR3 (Q6P9H4) (Met1-Val352) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-9433