Learn More
Sino Biological Recombinant Human MAN1A2 Protein (His Tag)
Supplier:  Sino Biological 15195H07H100
Encompass_Preferred
A DNA sequence encoding the human MAN1A2 ( NP_006690.1) (Pro58-Arg641) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9435