Learn More
Sino Biological Recombinant Human RBP7 Protein (His Tag)
Supplier:  Sino Biological 15214H07E100
Encompass_Preferred
A DNA sequence encoding the human RBP7 (NP_443192.1) (Met1-Ala134) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9441