Learn More
Sino Biological Recombinant Human GIP Protein (hFc Tag)
Supplier:  Sino Biological 15340H02H100
Encompass_Preferred
A DNA sequence encoding the human GIP (NP_004114.1) (Met1-Gln93) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9483