Learn More
Sino Biological Recombinant Human COASY Protein (His Tag)
Supplier:  Sino Biological 15397H07E100
Encompass_Preferred
A DNA sequence encoding the human COASY (Q13057-1) (Met296-Asp564) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9512