Learn More
Sino Biological Recombinant Human GMFG Protein (His Tag)
Supplier:  Sino Biological 15439H07E100
Encompass_Preferred
A DNA sequence encoding the human GMFG (O60234) (Met1-Arg142) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9534