Learn More
Sino Biological Recombinant Human LRRC15 Protein (ECD, hFc Tag)
Supplier:  Sino Biological 15786H02H100
Encompass_Preferred
A DNA sequence encoding the human LRRC15 (NP_570843.2) (Met1-Gly538) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9731