Learn More
Fluidigm Corp Maxpar X8 Antibody Labeling Kit, 158Gd—4 Rxn
SDP
Supplier:  Fluidigm Corp 201158A
Encompass
This Maxpar kit lets you conjugate IgG antibodies with the 158Gd metal isotope, with each kit containing enough material to conjugate 4-40 antibodies in 100 µg amounts.
Catalog No. NC0487814