Learn More
Chemglass Life Sciences Chemglass 15 L Bio Reactor Baffle Assembly, M10 mount
SDP
Supplier:  Chemglass Life Sciences CLS138517
Encompass
Chemglass 15L Bio Reactor Baffle Assembly (CLS-1385-17) is a stainless-steel baffle designed to improve mixing efficiency and reduce vortex formation in 15 L glass bioreactors. It mounts near the reactor wall using an M10 thread and is suitable for fermentation and cell culture applications.
  • Improves mixing efficiency and reduces vortexing.
  • 316L stainless steel construction.
  • Electropolished surface finish for corrosion resistance and cleanability.
  • M10 mounting for secure installation near the reactor wall.
  • Compatible with 15 L glass bioreactors.
  • Replacement O-ring available for routine maintenance.
Catalog No. 50-273-3306