Learn More
Chemglass Life Sciences Chemglass 15 mL Heavy-Wall Round-Bottom Flask, Single Neck, 14/20 Outer Joint with Hooks
SDP
Supplier:  Chemglass Life Sciences CG150705
Encompass_Preferred
15 mL heavy-wall single-neck round-bottom flask with a standard taper 14/20 outer joint and two hooks positioned 180° apart for secure spring support. Suitable for small-scale laboratory distillation, heating, and reflux applications.
  • Heavy-wall construction for improved durability.
  • Single short neck with a 14/20 outer taper joint.
  • Two hooks 180° apart for spring support.
  • 15 mL nominal volume for small-scale reactions.
  • Designed for use with stainless steel spring supports for secure mounting.
  • Suitable for distillation, heating, and reflux operations.
Catalog No. 50-273-3592