Learn More
Ace Glass, Inc. 15mL glass tapered tube with pouring lip
SDP
Supplier:  Ace Glass, Inc. 832516
Encompass_Preferred
15mL glass tapered tube with a pouring lip, for use with a glass pestle (50974568) as a tissue grinder. The conical area of the tube is for initial grinding, and the cylindrical area is for final homogenization. Interchangeably ground to 0.1-0.15mm clearance. Capacity represents volume with pestle inserted.
Catalog No. 50-979-772