Learn More
Ace Glass, Inc. 15mm adapter with drip tip 2mm bore with 1:5 taper PTFE stopcock
SDP
Supplier:  Ace Glass, Inc. 583511
Encompass_Preferred
15mm adapter with drip tip for chemical type chromatography. Adapter has 2mm bore with 1:5 taper PTFE stopcock at bottom for controlling flow.
Catalog No. 50-972-232