Learn More
Cole-Parmer Cole-Parmer PTFE Syringe Filters, Non-Sterile; 0.20 µm, 15 mm Diameter
Supplier:  Cole-Parmer 2955008
Encompass_Preferred
Filters are Triton-free and have a bidirectional membrane support and luer lock (15/25 mm dia) or stepped barbs (50 mm dia) inlet/outlet for a secure fit to the syringe. Membrane type and pore size are printed on each filter for easy identification. All Cole-Parmer syringe filters are 100% integrity tested and manufactured in accordance with ISO 9002 standards. Sterile filters are sterilized by gamma radiation. They are certified nonpyrogenic, noncytotoxic, and are blister packed.
Catalog No. NC9444017