Learn More
Cayman Chemical 15PGDH PosItIve CNTRLPGE 250uL
SDP
Supplier:  Cayman Chemical 701983250 UL
Encompass_Preferred
A styryl dye and an inhibitor of mechanosensitive ion channels reversibly binds to cell membranes and has been used to monitor synaptogenesis and reinnervation endocytosis and vacuole formation in vitro inhibits rapidly and slowly adapting mechanically activated currents in both capsaicin-insensitive and capsaicin-sensitive isolated rat dorsal root ganglion neurons from 0.6-15 UM ex/em 485/535 nm respectively
Catalog No. 50-289-2591