Learn More
Medchemexpress LLC 15R-Bimatoprost 1mg | 1163135-92-9 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1148671MG
Encompass_Preferred
(15R)-Bimatoprost is a prostaglandin analog and the 15-epi isomer of bimatoprost, supplied as a small-quantity analytical standard for research on glaucoma and ocular hypertension. It is used in laboratory studies, method development, and as a reference control for ophthalmic research.
  • Prostaglandin analog and 15-epi isomer of bimatoprost.
  • Intended for glaucoma and ocular hypertension research.
  • Suitable as a reference standard for analytical and method development applications.
  • Available in small pack sizes for laboratory use (e.g., 1 mg).
  • Documentation such as a data sheet and safety data sheet (SDS) available for specifications and handling.
Catalog No. 50-002-29519