Learn More
S2 Media 15x100mm Brucella Agar with 5% Sheep Blood, 10/pack
Supplier:  S2 Media 5396
Encompass_Preferred
S2Media™ Brucella Agar with 5% Sheep Blood is a non-selective and differential medium used in qualitative procedures for the cultivation of Brucella species and other fastidious microorganisms. The medium is based on the APHA Albimi broth formulation utilized for cultivation of fastidious microorganisms. The addition of 5% sheep blood enhances nutritional value and enable hemolysis detection. • Product meets APHA, ASM, CLSI, ISO, SM-Dairy, and USDA performance criteria.• Brucella Agar with 5% Sheep Blood should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5940